Oxymetazoline hydrochloride ≥99% (por HPLC), Sigma-Aldrich®
Fornecedor: Merck
|
Perigo
|
Sinônimos:
SOxymetazoline hydrochloride, 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrin hydrochloride
Nível de qualidade M-Clarity™ = MQ100
O2378-25G
O2378-5G
SIALO2378-25GEA
954
EUR
SIALO2378-25G
SIALO2378-5G
Oxymetazoline hydrochloride ≥99% (por HPLC), Sigma-Aldrich®
Oxymetazoline hydrochloride
Partial agonist.
Oxymetazoline hydrochloride is a partial α2A-adrenoceptor agonist; agonist at 5-HT1A, 5-HT1B, and 5-HT1D serotonin receptors. It is a mixed agonist-antagonist at 5-HT1C serotonin receptors.
Soluble in water (50 mg/ml) and alcohol.
Fórmula:
C₁₆H₂₄N₂O·HCl |
Número MDL :
MFCD00058147 Número CAS: 2315-02-8 EINECS: 219-015-0 UN: 2811 ADR: 6.1,I |
Especificações dos Resultados do Teste
Carbon | 63.8 - 65.6% |
Nitrogen | 9.1 - 9.7% |
Purity (HPLC) | ≥99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...