Print…
Lyticase (from Arthrobacter luteus)
Fornecedor: Merck
L4025-100KU L4025-1MU L4025-250KU L4025-25KU L4025-50KU
SIALL4025-100KUEA 552 EUR
SIALL4025-100KU SIALL4025-1MU SIALL4025-250KU SIALL4025-25KU SIALL4025-50KU
Lyticase (from Arthrobacter luteus)
Enzimas
Lyticase hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan.
  • Yeast cells are difficult to disrupt because the cell walls may form capsules or resistant spores. DNA can be extracted from yeast by using lysing enzymes such as lyticase, chitinase, zymolase, and gluculase to induce partial spheroplast formation.