Potassium methanesulphonate, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos:
Potassiummethanesulphonate, Methanesulphonic acid potassium salt
83000-25G-F
83000-5G-F
SIAL83000-25G-FEA
156
EUR
SIAL83000-25G-F
SIAL83000-5G-F
Potassium methanesulphonate, Sigma-Aldrich®
Potassium methanesulphonate
Fórmula:
CH₃KO₃S Peso Molecular: 134.2 g/mol |
Número MDL :
MFCD00070544 Número CAS: 2386-56-3 EINECS: 219-202-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...