Print…

Silver trifluoromethanesulphonate, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt

Nível de qualidade M-Clarity™ = MQ100

483346-1G 483346-5G
SIAL483346-1GEA 96.8 EUR
SIAL483346-1G SIAL483346-5G
Silver trifluoromethanesulphonate, Sigma-Aldrich®
Silver trifluoromethanesulphonate
Fórmula: CAgF₃O₃S
Peso Molecular: 256.94 g/mol
Ponto de Fusão: 356
Número MDL : MFCD00013226
Número CAS: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8 III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR