Print…

Nickel(II) chloride hydrate, Sigma-Aldrich®

Fornecedor: Merck
Perigo


Nível de qualidade M-Clarity™ = MQ100

364304-100G 364304-500G
SIAL364304-100GEA 99.3 EUR
SIAL364304-100G SIAL364304-500G
Nickel(II) chloride hydrate, Sigma-Aldrich®
Nickel(II) chloride hydrate
Fórmula: Cl₂Ni·H₂O
Peso Molecular: 147.61 g/mol
Número MDL : MFCD00149808
Número CAS: 69098-15-3
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR