Print…

Cloreto de guanidínio 99,5 Molecular_Biology

Fornecedor: Promega
Atenção

Sinônimos: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

H5381 H5383
PROMH53810100EA 89.3 EUR
PROMH53810100 PROMH5383
Cloreto de guanidínio 99,5 Molecular_Biology
Cloreto de guanidínio

Guanidine-HCl, Molecular Grade, is commonly used for the isolation of intact mRNA from tissues or cultured cells.


  • Quality tested and certified free of DNase, RNase and protease activity


Form: Fine, colorless or white crystals.
Storage: 15°C to 30°C

Fórmula: NH₂C(=NH)NH₂·HCl
Peso Molecular: 95.53 g/mol
Ponto de Fusão: 185
Densidade: 1.354 g/cm³ (20 °C)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00013026
Número CAS: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Purity ≥99.5%
A230 at 6M ≤0.15
A260 at 6M ≤0.03
A280 at 6M ≤0.02
Moisture ≤0.3%
Melting Point 186–188°C
Lead ≤5 ppm
Zinc ≤1 ppm
Copper ≤1ppm
Iron ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR