Print…
Transport boxes, Securipack
Fornecedor: E3 Cortex
FABRSECU1G1 FABRSECU03G1 FABRSECU05G1 FABRSECU2L5G1 FABRSECU5G1
CORTFABRSECU1G1EA 139 EUR
CORTFABRSECU1G1 CORTFABRSECU03G1 CORTFABRSECU05G1 CORTFABRSECU2L5G1 CORTFABRSECU5G1
Transport boxes, Securipack
Caixas Caixas de transporte
This Securipack packages are ready for use in the transport of samples of hazardous materials, except for Classes 7 and 6.2.
  • Very simple to use, this package eliminates risk posed by error or oversight
  • Mechanical high strength, in particular in the stacking by the juxtaposition of double walls on all the faces
  • High absorption power of the shocks on all the faces, guaranteeing a perfect protection in case of an accidental fall
Universal application for products regulated except for the radioactive and infectious products.
Protects against the following risks: It is a good oxidiser; non flammable and non toxic; corrosive
Informações de encomenda: Supplied with 1 case (sheath), 2 crossed envelopes dressed in a foam thickness, 1 pocket of metallic or plastic retention and 1 sluggish absorbing product. Interior packaging and hazard class label may be supplied upon request.