Print…

n-Decane-[D₂₂], Sigma-Aldrich®

Fornecedor: Merck
Perigo


Nível de qualidade M-Clarity™ = MQ200

452416-1G 452416-5G
SIAL452416-1GEA 342 EUR
SIAL452416-1G SIAL452416-5G
n-Decane-[D₂₂], Sigma-Aldrich®
n-Decane-[D₂₂]
Fórmula: D₃C(CD₂)₈CD₃
Número CAS: 16416-29-8
UN: 2247
ADR: 3,III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR