Print…

Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥94.0% estabilizada, líquido, Sigma-Aldrich®

Fornecedor: Merck
Perigo

Sinônimos: Acrylic Acid Hydroxypropyl Ester, Propylene Glycol Monoacrylate

Estabilização: 200 ppm Estabilizado com éter monometílico de hidroquinona (4-metoxifenol, MEHQ)
Nível de qualidade M-Clarity™ = MQ200

370932-18L 370932-1L
SIAL370932-18LEA 1120 EUR
SIAL370932-18L SIAL370932-1L
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥94.0% estabilizada, líquido, Sigma-Aldrich®
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate)

Hydroxypropyl acrylates may be used for the synthesis of a number of acrylates. The presence of both vinyl and hydroxyl allows copolymerization of the acrylate with vinyl monomers as well as with di- or polyisocyanates, anhydrides, or epoxides.


Hydroxypropyl acrylate, mixture of isomers contains 200 ppm hydroquinone monomethyl ether as inhibitor.

Fórmula: C₆H₁₀O₃
Peso Molecular: 130.14 g/mol
Ponto de ebulição: 77 °C (5 mmHg) (l)
Densidade: 1.044 g/ml (25 °C)
Ponto de Inflamação: 99 °C
Número MDL : MFCD04113589
Número CAS: 25584-83-2
UN: 1760
ADR: 8 III

Encomendar

Especificações dos Resultados do Teste

Colour Colourless
Form Liquid
Infrared spectrum Conforms to structure
Assay 95%
Mixture of isomers MEHQ as inhibitor 180 – 220 ppm
Vapor density (vs air) 4,5
Refractive index n20/D 1,445 (l)
Density (25 °C) 1,044 g/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR