Print…
Preparative sorbents, LiChroprep®
Fornecedor: MERCK PRODUCTION CHEMICALS
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
1.09303.0100EA 524 EUR
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
Preparative sorbents, LiChroprep®
Cromatografia, Fase estacionária HPLC, Fase Estacionária
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.