Print…
VWR® Water and General Sampling Bottles
Fornecedor: VWR Collection
331-0309
331-0305EA 146 EUR
331-0305 331-0309 VWRI331-0309.1 VWRI331-0306.1 VWRI331-0307.1 VWRI331-0308.1
VWR® Water and General Sampling Bottles
Frascos Frascos para amostras de água
HDPE, translucent, food grade wide neck graduated square bottles with HDPE colour coded tamper-evident caps.
  • Colour coded caps for easy identification
  • Tamper-evident screw caps for all bottle types
  • Fully batch traceable for irradiated items
  • Bottles individually numbered for better traceability
  • Fully recyclable
  • Square shape for easy storage and handling
  • Graduated at 100 ml increments
Lightweight, durable bottles, graduated. Choice of non sterile, sterile and sterile dosed with sodium thiosulphate. Wide neck (38 mm) for easy filling and emptying.