Print…
Human recombinant Angiopoietin-like 8 (from cells)
Fornecedor: ProSci
PRSI91-639EA 761 EUR
PRSI91-639
Human recombinant Angiopoietin-like 8 (from cells)
Proteínas e Peptídeos
The protein specifically promotes pancreatic beta cell proliferation and beta cell mass expansion, thereby improving glucose tolerance. It promotes pancreatic beta cell proliferation without insulin resistance. Also it acts as a blood lipid regulator by regulating serum triglyceride levels and possibly by promoting ANGPTL3 cleavage. It interacts with ANGPTL3. It predominantly expressed in liver and also expressed in adipose tissues. The ability of the protein to induce pancreatic beta cell proliferation is promising in diabetes therapy. Betatrophin treatment could supply or replace insulin injections by increasing the number of insulin-producing cells in diabetes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Fc tag