Print…
Fluorometer, Quantus®
Fornecedor: Promega
Quantus®
PROME6150EA 2590 EUR
PROME6150
Fluorometer, Quantus®
Spectroscopic Instruments Espectrofotómetros de fluorescência
Quantus is a compact, dual-channel fluorometer designed to provide highly sensitive fluorescent detection, when quantifying nucleic acids.
  • Ready to use with QuantiFluor® dyes
  • Recommended for next-generation sequencing (NGS) technology
  • Compatible with illumina (HiSeq/miSeq), Roche (454), as well as lifetech (ion Torrent and ion Proton) systems
Pre-programmed settings for QuantiFluor dye systems (QuantiFluor® dsDNA, RNA and ssDNA systems) to quantitate nucleic acids and flexibility to create customized methods and quantitation settings for other fluorescent dyes.
Operating system: Windows® 8, Windows 7 (32 or 64 bit) .NET Framework 4.0 or later at least one USB port available.