Ribonucleotide Triphosphates (rNTPs)
Fornecedor: Promega
E6000
E6011
E6021
E6031
E6041
P1132
P1142
P1152
P1162
PROME60001600EA
376
EUR
PROME60001600
PROME60110400
PROME60210400
PROME60310400
PROME60410400
PROMP11320500
PROMP11420500
PROMP11520500
PROMP11620500
Ribonucleotide Triphosphates (rNTPs)
Reagentes de ácidos nucleicos
Nucleótidos
Ribonucleotide triphosphates (rNTPs) compatible with the Riboprobe® and HeLaScribe® systems.
- Individual tubes containing 400 µl rNTP supplied at 10 mM or 100 mM in water at neutral pH
- Qualified for use with the Riboprobe® and HeLaScribe® systems
- Purity verified by HPLC
Fornecimento: Supplied in nuclease-free water.