Print…
Ribonucleotide Triphosphates (rNTPs)
Fornecedor: Promega
E6000 E6011 E6021 E6031 E6041 P1132 P1142 P1152 P1162
PROME60001600EA 376 EUR
PROME60001600 PROME60110400 PROME60210400 PROME60310400 PROME60410400 PROMP11320500 PROMP11420500 PROMP11520500 PROMP11620500
Ribonucleotide Triphosphates (rNTPs)
Reagentes de ácidos nucleicos Nucleótidos
Ribonucleotide triphosphates (rNTPs) compatible with the Riboprobe® and HeLaScribe® systems.
  • Individual tubes containing 400 µl rNTP supplied at 10 mM or 100 mM in water at neutral pH
  • Qualified for use with the Riboprobe® and HeLaScribe® systems
  • Purity verified by HPLC
Fornecimento: Supplied in nuclease-free water.