Genomic DNA, human
Fornecedor: Promega
G1471
G1521
G3041
PROMG14710100EA
303
EUR
PROMG14710100
PROMG15210100
PROMG30410100
Genomic DNA, human
Electroforese, Reagentes
Western Blotting Reagents
Human Genomic DNA is isolated from multiple anonymous donors.
Human Genomic DNA is suitable for southern blot hybridisations, genomic analysis (including PCR) and genomic library construction. Genomic DNA from selected species is purified and stored in 10 mM Tris-HCl (pH 8,0), 1 mM EDTA. Greater than 90% of the DNA is longer than 50 kb in size as measured by pulsed-field gel electrophoresis.