Print…
Genomic DNA, human
Fornecedor: Promega
G1471 G1521 G3041
PROMG14710100EA 303 EUR
PROMG14710100 PROMG15210100 PROMG30410100
Genomic DNA, human
Electroforese, Reagentes Western Blotting Reagents
Human Genomic DNA is isolated from multiple anonymous donors.
Human Genomic DNA is suitable for southern blot hybridisations, genomic analysis (including PCR) and genomic library construction. Genomic DNA from selected species is purified and stored in 10 mM Tris-HCl (pH 8,0), 1 mM EDTA. Greater than 90% of the DNA is longer than 50 kb in size as measured by pulsed-field gel electrophoresis.