Print…
dsDNA Quantification system, QuantiFluor® One
Fornecedor: Promega
E4871 E4870
PROME4871EA 167 EUR
PROME4871 PROME4870
dsDNA Quantification system, QuantiFluor® One
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The QuantiFluor® ONE dsDNA System provides a fluorescent double-stranded DNA-binding dye (504 nm Ex/531 nm Em) in an automation-friendly, "Add-and-Read" format for both dye and standard, simplifying DNA quantitation and speeding up workflow.
  • Specific quantitation of dsDNA
  • Increased sensitivity over absorbance at 260 nm for low-concentration samples
  • More sensitive for low-concentration samples than other available methods
Fornecimento: Kit includes QuantiFluor® ONE dsDNA dye (1×20 ml), Lambda DNA standard (1×80 µg) and 1X TE buffer, pH 7,5 (1×10 ml).