dsDNA Quantification system, QuantiFluor® One
Fornecedor: Promega
The QuantiFluor® ONE dsDNA System provides a fluorescent double-stranded DNA-binding dye (504 nm Ex/531 nm Em) in an automation-friendly, "Add-and-Read" format for both dye and standard, simplifying DNA quantitation and speeding up workflow.
- Specific quantitation of dsDNA
- Increased sensitivity over absorbance at 260 nm for low-concentration samples
- More sensitive for low-concentration samples than other available methods
Fornecimento: Kit includes QuantiFluor® ONE dsDNA dye (1×20 ml), Lambda DNA standard (1×80 µg) and 1X TE buffer, pH 7,5 (1×10 ml).