RNA purification, ReliaPrep™ FFPE Total RNA Miniprep System
Fornecedor: Promega
Z1001
Z1002
PROMZ1001EA
144
EUR
PROMZ1001
PROMZ1002
RNA purification, ReliaPrep™ FFPE Total RNA Miniprep System
Reagentes de ácidos nucleicos
Nucleic Acid Purification Kits and Reagents
The ReliaPrep™ FFPE Total RNA Miniprep System optimises lysis conditions to reverse RNA modifications that can be introduced when extracting RNA from FFPE tissues.
- No need for overnight digestion
- Safely deparaffinise without harsh organic solvents
- Elimination of the washing steps involved in xylene deparaffinisation means small RNA fragments are retained
Fornecimento: Kit includes enough mineral oil; lysis buffer (LBA); proteinase K (PK) solution; MnCl2 (0.09 M); BL buffer; wash solution; DNase I (lyophilised); DNase buffer; nuclease-free water; collection tubes; ReliaPrep™ FFPE binding columns; and elution tubes to perform total rRNA isolations.