Print…

Anti-GLB1 Mouse Monoclonal Antibody

Fornecedor: Promega
Z3781 Z3783
PROMZ3781EA 80.9 EUR
PROMZ3781 PROMZ37832000
Anti-GLB1 Mouse Monoclonal Antibody
Anticorpos
Antibody [subclass IgG2a(κ)] was purified from ascites of a mouse hybridoma and recognises E. coli β-galactosidase near the C-terminal end.

  • Antibody is supplied as 2.0 - 2.5 mg/ml in 10 mM Tris-HCl (pH 8.0), 150 mM NaCl, 0.02% sodium azide.

Type:
Antigen: ß-Galactosidase
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR