Trypsin/Lys-C mix, MS grade
Fornecedor: Promega
V5072
V5071
PROMV5072EA
282
EUR
PROMV5072
PROMV5071
Trypsin/Lys-C mix, MS grade
Extracção de proteínas
Reagentes de Extração de Proteínas
Trypsin/Lys-C mix improves peptide digestion over trypsin alone, increasing peptide recovery and resulting in better protein quantitation by using standard digestion conditions.
- Improve protein digestion with mix of Trypsin Gold and rLys-CUse standard overnight digestion, non denaturing conditions
- Increased peptide recovery, enhanced protein quantitation, improved reproducibility
- Tolerant to trypsin-inhibiting contaminants
- Suppress artificial non enzymatic post translational modification without added digestion time
Fornecimento: Kit includes Trypsin/Lys-C mix (mass spectrometry grade) and resuspension buffer.