Print…
Trypsin/Lys-C mix, MS grade
Fornecedor: Promega
V5072 V5071
PROMV5072EA 282 EUR
PROMV5072 PROMV5071
Trypsin/Lys-C mix, MS grade
Extracção de proteínas Reagentes de Extração de Proteínas
Trypsin/Lys-C mix improves peptide digestion over trypsin alone, increasing peptide recovery and resulting in better protein quantitation by using standard digestion conditions.
  • Improve protein digestion with mix of Trypsin Gold and rLys-CUse standard overnight digestion, non denaturing conditions
  • Increased peptide recovery, enhanced protein quantitation, improved reproducibility
  • Tolerant to trypsin-inhibiting contaminants
  • Suppress artificial non enzymatic post translational modification without added digestion time
Fornecimento: Kit includes Trypsin/Lys-C mix (mass spectrometry grade) and resuspension buffer.