Signaling pathway analysis response element firefly luciferase vectors
Fornecedor: Promega
E8471
E8491
E8481
E1340
E1350
E4611
PROME8471EA
762
EUR
PROME8471
PROME8491
PROME8481
PROME1340
PROME1350
PROME4611
Signaling pathway analysis response element firefly luciferase vectors
Vectors, Plasmids and Libraries
Ready-to-use reporter vectors used to analyse signaling pathway without cloning.
- Response elements are involved in nuclear response, kinase and other signaling pathways
- Each vector included an optimised firefly luciferase gene that codes a reporter with a one-hour half-life
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
These vectors are applicable for transcription regulation, virus-cell interactions, compound screening, post-translational modification, and GPCR and cell signalling.