Print…
Signaling pathway analysis response element firefly luciferase vectors
Fornecedor: Promega
E8471 E8491 E8481 E1340 E1350 E4611
PROME8471EA 762 EUR
PROME8471 PROME8491 PROME8481 PROME1340 PROME1350 PROME4611
Signaling pathway analysis response element firefly luciferase vectors
Vectors, Plasmids and Libraries
Ready-to-use reporter vectors used to analyse signaling pathway without cloning.
  • Response elements are involved in nuclear response, kinase and other signaling pathways
  • Each vector included an optimised firefly luciferase gene that codes a reporter with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
These vectors are applicable for transcription regulation, virus-cell interactions, compound screening, post-translational modification, and GPCR and cell signalling.