Print…
RNA isolation system, SV total RNA
Fornecedor: Promega
Z3100 Z3101 Z3105
PROMZ31000050EA 375 EUR
PROMZ31000050 PROMZ31010010 PROMZ3105
RNA isolation system, SV total RNA
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The SV Total RNA Isolation System provides a fast and simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.
  • Rapid isolation of high yields of total RNA without the use of hazardous compounds
  • Choice of microcentrifugation (spin) or vacuum purification methods
  • Purified RNA suitable for all routine molecular biology applications, including RT-PCR
SV Total RNA Isolation System incorporates a DNase treatment step directly on the minicolumn membrane. This step substantially reduces genomic DNA contamination, which can interfere with amplification-based methodologies. Purification is achieved without the use of phenol:chloroform extractions or ethanol precipitations, and there is no DNase carry over in the final RNA preparation.
Fornecimento: Kit includes collection tubes; elution tubes; spin columns; RNA lysis buffer (RLA); RNA dilution buffer (RDA) (blue buffer); β-mercaptoethanol (97,4%); DNase I (lyophilised); MnCl₂ (0,09 M); yellow core buffer; DNase stop solution (DSA) (concentrated); RNA wash solution (RWA) (concentrated); nuclease-free water.