Print…
Human protein extracts (intact)
Fornecedor: Promega
PROMV6941EA 235 EUR
PROMV6941
Human protein extracts (intact)
Proteínas e Peptídeos
These whole-cell protein extract prepared from human K562 cells are designed for monitoring mass spec instrument performance.
  • Extract reproducibility is assured by stringent control over the manufacturing process
  • Each protein extract is monitored by LC-MS to confirm composition and protein integrity
  • To be used as test material for optimizing mass spec protein sample preparation
The extracts are predigested and cleaned up by solid-phase extraction for immediate use in liquid chromatography/mass spectrometry (LC-MS) analysis.