Print…
In Vivo Renilla Luciferase substrate, ViviRen™
Fornecedor: Promega
P1231 P1232
PROMP1231EA 287 EUR
PROMP1231 PROMP1232
In Vivo Renilla Luciferase substrate, ViviRen™
Sistemas de ligação e rotulagem de proteínas
ViviRen™ in vivo Renilla luciferase substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimise substrate degradation and autoluminescence. ViviRen™ Substrate is reported to demonstrate brighter output compared to the native coelenterazine substrate when used in an in vivo imaging application in a mouse model. High-quality substrate eliminates potential interference in assays due to the presence of endotoxins.
Embalagem: Product integrity is ensured by packaging most products in amber vials with septa and packaged with fine tolerances to minimise the need to weigh substrates.