Print…
Magnetic DNA purification kits for food, Wizard®
Fornecedor: Promega
FF3750 FF3751
PROMFF3750EA 1360 EUR
PROMFF3750 PROMFF3751
Magnetic DNA purification kits for food, Wizard®
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The Wizard® Magnetic DNA Purification Kits for Food are used to isolate DNA from a variety of food samples like corn seeds, cornmeal, soybeans, soy flour and soy milk. Processed food, such as corn chips, chocolate and chocolate-containing foods, lecithin and vegetable oils can also be used with protocol variations.
  • Requires minimal centrifugation and eliminates organic extractions
  • Purified DNA suitable for PCR-based GMO testing
  • Generates results in one-third of the time of traditional methods
The Wizard® Magnetic DNA Purification Kit for Food uses paramagnetic particles for DNA isolation. Unlike column-based systems, binding of nucleic acids to magnetic particles can occur in solution, resulting in increased binding kinetics and efficiency. Particles also can be completely resuspended during the wash steps of a purification protocol, thus enhancing contact with and removal of contaminants, and increasing nucleic acid purity.