CellTiter 96® AQueous MTS Reagent Powder
Fornecedor: Promega
G1111
G1112
PROMG11111000EA
1340
EUR
PROMG11111000
PROMG11120250
CellTiter 96® AQueous MTS Reagent Powder
Ensaios
Ensaios celulares
CellTiter 96® AQueous MTS Reagent Powder is a novel tetrazolium compound used in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays.