Print…
CellTiter 96® AQueous MTS Reagent Powder
Fornecedor: Promega
G1111 G1112
PROMG11111000EA 1340 EUR
PROMG11111000 PROMG11120250
CellTiter 96® AQueous MTS Reagent Powder
Ensaios Ensaios celulares
CellTiter 96® AQueous MTS Reagent Powder is a novel tetrazolium compound used in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays.