Restriction enzymes, PvuII
Fornecedor: Promega
R6331
R6335
PROMR63311000EA
0
EUR
PROMR63311000
PROMR63355000
Restriction enzymes, PvuII
Reagentes de ácidos nucleicos
Restriction Enzymes
These PvuII restriction enzyme recognises the cut site CAG˅CTG and GTC˄GAC. Star activity may be observed with glycerol concentrations >12% or with enzyme: DNA ratios >25 U/μg.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Incubation with buffer B at 37 °C
- Source: Proteus vulgaris
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: 1×1 ml buffer B 10X, 1×150 µl BSA, acetylated with concentration of 10 mg/ml, 1×1,000 U PvuII and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...