Restriction enzymes, PvuI
Fornecedor: Promega
R6321
R6325
PROMR63210100EA
120
EUR
PROMR63210100
PROMR63250500
Restriction enzymes, PvuI
Reagentes de ácidos nucleicos
Restriction Enzymes
These PvuI restriction enzyme recognises the cut site of CG AT˅CG and GC˄TA GC. Ends generated by PvuI can be directly ligated to ends generated by SgfI. The PvuI site is regenerated in the ligation product, but the SgfI site is not.
- Relegation of DNA fragments with dam methylation is difficult
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Incubation with buffer D at 37 °C
- Source: Proteus vulgaris
Storage Buffer: 10 mM Tris-HCl (pH 7,3 @ 25 °C), 50 mM NaCl, 0,1% Triton® X-100, 1 mM DTT, 0,1 mM EDTA, 0,5 mg/ml BSA and 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer D 10X, 1×150 µl BSA acetylated with concentration of 10 mg/ml and 1×100 U PvuI.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...