Restriction enzymes, DdeI
Fornecedor: Promega
R6291
R6295
PROMR62910200EA
0
EUR
PROMR62910200
PROMR62951000
Restriction enzymes, DdeI
Reagentes de ácidos nucleicos
Restriction Enzymes
These DdeI restriction enzyme recognises the cut site C˅TNA G and G ANT˄C. Produces an ambiguous extension, which is difficult to ligate.
- Capable of digesting DNA in 15 minutes or less
- Incubation with buffer D at 37 °C
- Source: Desulfovibrio desulfuricans
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 0,01% Triton® X-100 and 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer D 10X, 1×150 µl BSA, acetylated with concentration of 10 mg/ml, 1×200 U DdeI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...