Print…
Transfection reagent, ViaFect™
Fornecedor: Promega
E4981 E4982
PROME4981EA 440 EUR
PROME4981 PROME4982
Transfection reagent, ViaFect™
Reagentes para transfecção
ViaFect™ transfection reagent is designed to transfect DNA into a wide variety of cell lines with high efficiency and low toxicity. ViaFect™ Transfection Reagent has been shown to transfect cell lines that are traditionally thought of as difficult to transfect (such as hematopoietic, primary and iPSC stem cell lines) with relatively high efficiency. The protocol does not require removal of serum or culture medium and does not require washing or changing of medium after introducing the reagent/DNA complex.
  • Simple protocol with minimal optimisation
  • Keeps the cells viable
  • Does not contain any animal-derived components
ViaFect Transfection Reagent is a novel formulation reagent designed to efficiently introduce DNA into a wide variety of cell lines. ViaFect Transfection Reagent has performed in commonly used adherent cell models and also in cells lines traditionally thought of as difficult to transfect such as suspension cells and stem cell-derived lines. This gentle, low-toxicity reagent allows cells to stay healthy and metabolically active during transfection-based experiments and offers an easy-to-use protocol that does not require removal of serum or culture medium prior to use. After introducing the reagent:DNA complex no washing or medium changes are required. ViaFect Transfection Reagent provides robust performance with minimal optimization allowing simple design of more relevant assays.