Print…
Luciferase assay system, ONE-Glo™
Fornecedor: Promega
E6110 E6120
PROME6110EA 248 EUR
PROME6110 PROME6120
Luciferase assay system, ONE-Glo™
Ensaios Ensaios celulares
The ONE-Glo™ Luciferase Assay System provides a highly sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells in 96- and 384-well plates.
  • Improved assay precision: Less sensitive to mixing and dispensing conditions, enhancing reproducibility
  • Brighter and longer-lasting signal: Optimized for batch and continuous-process handling
  • Less sensitive to culture media, phenol red and luciferase inhibitors than other luciferase assays
Suitable for high- and ultrahigh-throughput applications, the ONE-Glo™ Assay contains a new luciferase substrate, resulting in a reagent that is more stable, more tolerant to sample components, and has less odor than standard luciferase assay reagents. These features ensure that the ONE-Glo™ Assay provides robust performance and also eliminates many of the handling inconveniences experienced using other reporter assays in a high-throughput setting.