Print…
Transfection reagent, FuGENE® 6
Fornecedor: Promega
E2692 E2691 E2693
E2692EA 1970 EUR
E2692 E2691 E2693
Transfection reagent, FuGENE® 6
Reagentes para transfecção
FuGENE® 6 transfection reagent is a non-liposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity.
  • Preserve cell viability
  • Straightforward protocol and minimal optimisation
  • Ideal for use with luciferase reporter gene assays: More expression; sensitive results
  • Simple protocol eliminates tedious culture medium changes and eliminates a source of variation
FuGENE is a registered trademark of Fugent, LLC, USA.