Transfection reagent, FuGENE® 6
Fornecedor: Promega
E2692
E2691
E2693
E2692EA
1970
EUR
E2692
E2691
E2693
Transfection reagent, FuGENE® 6
Reagentes para transfecção
FuGENE® 6 transfection reagent is a non-liposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity.
- Preserve cell viability
- Straightforward protocol and minimal optimisation
- Ideal for use with luciferase reporter gene assays: More expression; sensitive results
- Simple protocol eliminates tedious culture medium changes and eliminates a source of variation
FuGENE is a registered trademark of Fugent, LLC, USA.