Ribonuclease
Fornecedor: Promega
RNase ONE™ Ribonuclease is a 27 kDa periplasmic enzyme from E. coli that catalyses the degradation of RNA to cyclic nucleotide monophosphate (NMP) intermediates. Slower hydrolysis further catalyses the degradation of these intermediates to 3´-NMPs. RNase ONE™ Ribonuclease may be used to remove RNA from DNA preparations, for mismatch detection, RNase protection assays and mapping, or quantitation of RNA by selective cleavage of single-stranded regions.
- Able to cleave a phosphodiester bond between any two ribonucleotides
Fornecimento: Kit includes RNase ONE™ Ribonuclease (5 to 10 U/µl) and RNase ONE™ 10X Reaction Buffer (100 mM Tris-HCl, pH 7,5 at 25 °C; 50 mM EDTA; 2M sodium acetate).