dNTP sets (dATP, dCTP, dGTP, dUTP)
Fornecedor: Promega
U1245
U1335
PROMU1245EA
338
EUR
PROMU1245
PROMU1335
dNTP sets (dATP, dCTP, dGTP, dUTP)
Reagentes de ácidos nucleicos
Nucleótidos
High-quality deoxynucleotide triphosphates (dNTPs) are important for many key procedures such as cDNA synthesis, sequencing, cloning and genotyping.
- Highly pure (≥99% triphosphate)
- Each dNTP supplied at 100 mM in water for ease of use
dUTP (2'-Deoxyuridine, 5'-Triphosphate) can be used in place of dTTP in PCR and RT-PCR protocols to prevent carryover from previous amplifications. The substitution of dUTP for dTTP in PCR results in uracil-containing PCR products that are suitable for most standard applications.