Print…
dNTP sets (dATP, dCTP, dGTP, dUTP)
Fornecedor: Promega
U1245 U1335
PROMU1245EA 338 EUR
PROMU1245 PROMU1335
dNTP sets (dATP, dCTP, dGTP, dUTP)
Reagentes de ácidos nucleicos Nucleótidos
High-quality deoxynucleotide triphosphates (dNTPs) are important for many key procedures such as cDNA synthesis, sequencing, cloning and genotyping.
  • Highly pure (≥99% triphosphate)
  • Each dNTP supplied at 100 mM in water for ease of use
dUTP (2'-Deoxyuridine, 5'-Triphosphate) can be used in place of dTTP in PCR and RT-PCR protocols to prevent carryover from previous amplifications. The substitution of dUTP for dTTP in PCR results in uracil-containing PCR products that are suitable for most standard applications.