Print…
dNTP sets (dATP, dCTP, dGTP, dTTP)
Fornecedor: Promega
U1240 U1330 U1410 U1420
PROMU12400040EA 276 EUR
PROMU12400040 PROMU13300010 PROMU1410 PROMU14200025
dNTP sets (dATP, dCTP, dGTP, dTTP)
Reagentes de ácidos nucleicos Nucleótidos
High-quality deoxynucleotide triphosphates (dNTPs) are important for key procedures including cDNA synthesis, sequencing, cloning, and preparation of libraries for next generation sequencing.
  • PCR-tested deoxynucleotides ensure optimal performance with amplification enzymes
  • dNTPs are >99% pure, allowing highly consistent results
  • Supplied at a convenience concentration (100 mM) in water at pH 7,5 for versatile use in PCR and other applications