dNTP sets (dATP, dCTP, dGTP, dTTP)
Fornecedor: Promega
U1240
U1330
U1410
U1420
PROMU12400040EA
276
EUR
PROMU12400040
PROMU13300010
PROMU1410
PROMU14200025
dNTP sets (dATP, dCTP, dGTP, dTTP)
Reagentes de ácidos nucleicos
Nucleótidos
High-quality deoxynucleotide triphosphates (dNTPs) are important for key procedures including cDNA synthesis, sequencing, cloning, and preparation of libraries for next generation sequencing.
- PCR-tested deoxynucleotides ensure optimal performance with amplification enzymes
- dNTPs are >99% pure, allowing highly consistent results
- Supplied at a convenience concentration (100 mM) in water at pH 7,5 for versatile use in PCR and other applications