Restriction enzymes, EcoRV
Fornecedor: Promega
R6351
R6355
PROMR63512000EA
85.1
EUR
PROMR63512000
PROMR635510KU
Restriction enzymes, EcoRV
Reagentes de ácidos nucleicos
Restriction Enzymes
These EcoRV restriction enzyme recognises and cut GAT˅ATC and CTA^TAG at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Highly concentrated
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Source: Escherichia coli J62 pLG74.
Fornecimento: Kit includes 1×1 ml buffer D 10X buffer, 1×150 µl bovine serum albumin (acetylated), 1×2000 units EcoRV and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...