Restriction enzymes, KpnI
Fornecedor: Promega
R6341
R6345
PROMR63412500EA
108
EUR
PROMR63412500
PROMR634510KU
Restriction enzymes, KpnI
Reagentes de ácidos nucleicos
Restriction Enzymes
These KpnI restriction enzyme recognises G GTAC˅C and C^CATG G sites and cuts best at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Highly concentrated
Storage buffer: 10 mM Tris-HCl (pH 7,4), 200 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Source: Recombinant E. coli strain.
Fornecimento: Kit includes 1×1ml buffer J 10X buffer, 1×150 µl bovine serum albumin (acetylated), 1×2500 units KpnI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...