Print…
Luciferase SP6/T7 control DNAs
Fornecedor: Promega
L4741 L4821
PROML47410020EA 276 EUR
PROML47410020 PROML48210020
Luciferase SP6/T7 control DNAs
Expressão de proteínas Reagentes de Expressão de Proteínas
The Luciferase SP6 and T7 Control DNAs are used as functional controls in the TNT® Quick Coupled and TNT® Coupled transcription/translation systems.
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay
The control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter. All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are monitored easily by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components.
Fornecimento: Luciferase control DNAs are supplied as 0,5 mg/ml solutions in TE buffer.