pUC/M13 sequencing primers
Fornecedor: Promega
Q5401
Q5421
Q5601
PROMQ54010002EA
160
EUR
PROMQ54010002
PROMQ54210002
PROMQ56010002
pUC/M13 sequencing primers
Reagentes de ácidos nucleicos
Reagentes de Sequenciação
The pUC/M13 primers are used to sequence inserts cloned into the M13 vectors and pUC plasmids developed by Messing. These primers also can be used for sequencing other lacZ-containing plasmids such as the pGEM®-Z and pGEM®-Zf Vectors. The primers are purified by gel electrophoresis or HPLC.
Primer sequences:
Forward (17mer): 5´-d(GTTTTCCCAGTCACGAC)-3´
Reverse (17mer): 5´-d(CAGGAAACAGCTATGAC)-3´
Reverse (22mer): 5´-d(TCACACAGGAAACAGCTATGAC)-3´
Forward (24mer): 5´-d(CGCCAGGGTTTTCCCAGTCACGAC)-3´
Storage: –20 °C