DNA purification systems, Wizard® Magnetic 96 DNA Plant System
Fornecedor: Promega
The Wizard® Magnetic 96 DNA Plant System is designed for manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue. The system has been validated with corn and tomato leaf, as well as with canola and sunflower seeds. The DNA purified from these samples can be used in PCR as well as more demanding applications such as RAPD analysis. Unlike column-based systems, the binding of nucleic acids to magnetic particles can occur in solution, enhancing contact with the wash buffer and increasing nucleic acid purity.
- Cuts purification time compared to CTAB extraction
- Eliminates organic extractions
- Validated for both leaf and seed tissue by PCR and RAPD analysis
Informações de encomenda: Kit components can be ordered separately.
Fornecimento: The complete kit includes Plant Wash Buffer, collection plates (4 per pack), Lysing Buffer A and B, and MagneSil® paramagnetic particles.