Restriction enzymes, XbaI
Fornecedor: Promega
R6181
R6185
PROMR61812000EA
109
EUR
PROMR61812000
PROMR618510KU
Restriction enzymes, XbaI
Reagentes de ácidos nucleicos
Restriction Enzymes
The Xbal restriction enzyme is capable of digesting DNA in 15 minutes or less. It recognises T˅CTAG A and A GATC˄T. Genome qualified to ensure optimal performance in genomic analysis applications.
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue or white cloning qualified, providing a higher level of quality control for enzymes
- Incubation condition at 37 °C in buffer D
- Source: Xanthomonas badrii
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 300 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: The kit includes 1×1ml buffer D 10X buffer, 1×150 µl bovine serum albumin, acetylated with concentration of 10 mg/ml, 1×2,000 units XbaI and 1×0,25ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...