Restriction enzymes, HaeIII
Fornecedor: Promega
R6171
R6175
PROMR61712500EA
0
EUR
PROMR61712500
PROMR617510KU
Restriction enzymes, HaeIII
Reagentes de ácidos nucleicos
Restriction Enzymes
The HaeIII restriction enzyme is capable of digesting DNA in 15 minutes or less. It recognises the cut site GG˅CC and CC˄GG.
- Incubation at 37 °C in buffer C
- Source: Haemophilus aegyptius
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 300 mM NaCl, 0,1 mM EDTA, 1mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer C 10X buffer, 1×150 µl bovine serum albumin, acetylated with concentration of 10 mg/ml, 1×2,500 units HaeIII and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...