Restriction enzymes, XhoI
Fornecedor: Promega
R6161
R6165
PROMR61613000EA
84
EUR
PROMR61613000
PROMR616510KU
Restriction enzymes, XhoI
Reagentes de ácidos nucleicos
Restriction Enzymes
The XhoI restriction enzymes are capable of digesting DNA in 15 minutes or less. It recognises the cut site C˅TCGA G and G AGCT˄C. Genome qualified to ensure optimal performance in genomic analysis applications
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue or white cloning qualified, providing a higher level of quality control for enzymes
- Incubation at 37 °C with buffer D
- Source: Xanthomonas holcicola
- Highly concentrated
Storage Buffer: 10 mM Tris-HCl (pH 73), 300 mM KCl, 0,1% Triton® X-100, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer D 10X buffer, 1×150 µl bovine serum albumin, acetylated with concentration of 10 mg/ml and 1×3,000 units of XhoI.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...