Print…

D(-)-Luciferin potassium salt, VivoGlo™, grau in vivo

Fornecedor: Promega

Sinônimos: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate

P1041 P1042 P1043
PROMP1041EA 648 EUR
PROMP1041 PROMP1042 PROMP1043
D(-)-Luciferin potassium salt, VivoGlo™, grau in vivo
D-(-)-Luciferin potassium salt

Firefly Luciferase Substrate for Use in Animal Imaging Studies.


  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings


Luciferase genes from the North American firefly (Photinus pyralis) and from other beetles are commonly used as light-emitting reporters in cellular and animal models. VivoGlo™ Luciferin is the potassium salt of d-luciferin, the firefly luciferase substrate capable of generating light when a suitable model is used. Product integrity is ensured by packaging most products in amber vials with septa as well as offering ease of dilution and use for imaging experiments. VivoGlo™ Luciferin is packaged with fine tolerances to minimize the need to weigh substrates.
Storage: -30°C to -10°C

Fórmula: C₁₁H₇KN₂O₃S₂
Número MDL : MFCD00044928
Número CAS: 115144-35-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR