Restriction enzymes, TaqI
Fornecedor: Promega
R6151
R6155
PROMR61511000EA
98.7
EUR
PROMR61511000
PROMR615510KU
Restriction enzymes, TaqI
Reagentes de ácidos nucleicos
Restriction Enzymes
The TaqI restriction enzyme recognises the cut site T˅CG A and A GC˄T.
- Sensitive to overlapping dam methylation
- Exhibits <10% activity at 37 °C, with incubation condition at 65 °C in buffer E
- Source: Thermus aquaticus YTI
- Highly concentrated
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 300 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer E 10X buffer, 1×150 µl bovine serum albumin, acetylated with a concentration of 10 mg/ml, 1×1,000 units TaqI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...