Restriction enzymes, SmaI
Fornecedor: Promega
R6121
R6125
PROMR61211000EA
118
EUR
PROMR61211000
PROMR61255000
Restriction enzymes, SmaI
Reagentes de ácidos nucleicos
Restriction Enzymes
These SmaI restriction enzymes are genome qualified to ensure optimal performance in genomic analysis applications. Recognises CCC˅GGG and GGG˄CCC.
- Exhibits 25 - 50% activity at 37 °C with incubation condition of 25°C in buffer J
- Blue or white cloning qualified, providing a higher level of quality control for enzymes
- Source: Serratia marcescens
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 300 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: The kit includes 1×1 ml buffer J 10X buffer, 1×150 µl with concentration of 10 mg/ml, 1×1,000 units SmaI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...