PCR master mix (2X)
Fornecedor: Promega
M7501
M7502
M7505
PROMM7501AMEA
0
EUR
PROMM7501AM
PROMM7502
PROMM7505
PCR master mix (2X)
Reagentes de ácidos nucleicos
End-point PCR Enzymes and Kits
PCR Master Mix is a ready-to-use 2X solution containing Taq DNA polymerase, dNTPs, MgCl₂ and reaction buffers at optimal concentrations for efficient PCR amplification of DNA templates. The PCR Master Mix is designed for routine endpoint PCR for DNA amplicons in the range of 0,2 to 2 kb.
- Set up reactions with as low as 10 μl volume
- Optimized conditions enable amplification of as few as two copies of target template
- Stable for 3 months when stored at 4 °C
Fornecimento: Kit includes 2X PCR Master Mix and nuclease-free water.