Print…

Restriction enzymes, SalI

Fornecedor: Promega

R6051 R6055
PROMR60512000EA 155 EUR
PROMR60512000 PROMR605510KU
Restriction enzymes, SalI
Reagentes de ácidos nucleicos Restriction Enzymes

These SalI restriction enzyme recognises and cut G˅TCGA C and C AGCT^G at 37 °C in its own unique buffer.


  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq® Green master mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
  • Highly concentrated


Storage buffer: 10 mM Tris-HCl (pH 7,4), 300 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.


Source: Streptomyces albus G


Fornecimento: Kit includes 1×1 ml buffer D 10X buffer, 1×750 µl bovine serum albumin (acetylated) and 1×10 000 units SalI.

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR