Restriction enzymes, HincII
Fornecedor: Promega
R6031
R6035
R6037
PROMR60310200EA
0
EUR
PROMR60310200
PROMR60351000
PROMR60375000
Restriction enzymes, HincII
Reagentes de ácidos nucleicos
Restriction Enzymes
These HincII restriction enzyme recognises and cut GT(T/C)˅(A/G)AC and CA(A/G)^(T/C)TG at 37 °C in its own unique buffer.
- Active and capable of digestion directly in GoTaq® Green master mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Source: Haemophilus influenzae Rc
Storage Buffer: 10 mM Tris-HCl (pH 7.4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: Kit includes 1×1 ml buffer B 10X buffer, 1×750 µl bovine serum albumin (acetylated), 1×5,000 units HincII and 1×1 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...