Print…
Fluorescent ligands, HaloTag®
Fornecedor: Promega
G8581 G8591 G8251 G1001 G2801 G8272 G8281 G1002 G2802 G2991 G3221 G8252 G8273 G8282 G8582 G8592 G8471 G8472
PROMG8581EA 874 EUR
PROMG8581 PROMG8591 PROMG8251 PROMG1001 PROMG2801 PROMG8272 PROMG8281 PROMG1002 PROMG2802 PROMG2991 PROMG3221 PROMG8252 PROMG8273 PROMG8282 PROMG8582 PROMG8592 PROMG8471 PROMG8472
Fluorescent ligands, HaloTag®
Sistemas de ligação e rotulagem de proteínas
HaloTag® fluorescent ligands form covalent bonds rapidly under general physiological conditions. The bonds are highly specific and essentially irreversible. HaloTag® fluorescent ligands enable the addition of the chloroalkane group, with which HaloTag® protein reacts, to any compound or surface with a compatible chemical group.
Cell permeant TMR, Oregon Green®, diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins. Cell impermeant tags include Alexa Fluor® 488 and 660 for quick cell surface labeling.