Fluorescent ligands, HaloTag®
Fornecedor: Promega
G8581
G8591
G8251
G1001
G2801
G8272
G8281
G1002
G2802
G2991
G3221
G8252
G8273
G8282
G8582
G8592
G8471
G8472
PROMG8581EA
874
EUR
PROMG8581
PROMG8591
PROMG8251
PROMG1001
PROMG2801
PROMG8272
PROMG8281
PROMG1002
PROMG2802
PROMG2991
PROMG3221
PROMG8252
PROMG8273
PROMG8282
PROMG8582
PROMG8592
PROMG8471
PROMG8472
Fluorescent ligands, HaloTag®
Sistemas de ligação e rotulagem de proteínas
HaloTag® fluorescent ligands form covalent bonds rapidly under general physiological conditions. The bonds are highly specific and essentially irreversible. HaloTag® fluorescent ligands enable the addition of the chloroalkane group, with which HaloTag® protein reacts, to any compound or surface with a compatible chemical group.
Cell permeant TMR, Oregon Green®, diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins. Cell impermeant tags include Alexa Fluor® 488 and 660 for quick cell surface labeling.